missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(2,3-dihydro-1,4-benzodioxin-6-yl)ethan-1-one, Thermo Scientific™

$105.26 - $105.26

Chemical Identifiers

CAS 4629-54-3
Molecular Formula C10H9BrO3
Molecular Weight (g/mol) 257.08
MDL Number MFCD00099402
InChI Key CSSHRKYOZTZFCX-UHFFFAOYSA-N
Synonym 2-bromo-1-2,3-dihydro-1,4-benzodioxin-6-yl ethanone, 2-bromo-1-2,3-dihydro-1,4-benzodioxin-6-yl ethan-1-one, 2-bromo-1-2,3-dihydrobenzo b 1,4 dioxin-6-yl ethanone, 2-bromo-1-2,3-dihydrobenzo 1,4 dioxin-6-yl ethanone, 2-bromo-1-2,3-dihydrobenzo 1,4-dioxin-6-yl ethanone, 1,4-benzodioxan-6-ylbromomethyl ketone, 2-bromo-1-2,3-dihydro-benzo 1,4 dioxin-6-yl-ethanone, ethanone,2-bromo-1-2,3-dihydro-1,4-benzodioxin-6-yl
PubChem CID 2776171
SMILES BrCC(=O)C1=CC=C2OCCOC2=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC01305DA
SDS View Documents
Thermo Scientific Chemicals
CC01305DA
1 g
Each for $105.26
Only null left
 
Description

Description

Specifications

Chemical Identifiers

4629-54-3
257.08
CSSHRKYOZTZFCX-UHFFFAOYSA-N
2776171
C10H9BrO3
MFCD00099402
2-bromo-1-2,3-dihydro-1,4-benzodioxin-6-yl ethanone, 2-bromo-1-2,3-dihydro-1,4-benzodioxin-6-yl ethan-1-one, 2-bromo-1-2,3-dihydrobenzo b 1,4 dioxin-6-yl ethanone, 2-bromo-1-2,3-dihydrobenzo 1,4 dioxin-6-yl ethanone, 2-bromo-1-2,3-dihydrobenzo 1,4-dioxin-6-yl ethanone, 1,4-benzodioxan-6-ylbromomethyl ketone, 2-bromo-1-2,3-dihydro-benzo 1,4 dioxin-6-yl-ethanone, ethanone,2-bromo-1-2,3-dihydro-1,4-benzodioxin-6-yl
BrCC(=O)C1=CC=C2OCCOC2=C1

Specifications

4629-54-3
Amber Glass Bottle
MFCD00099402
2-bromo-1-2,3-dihydro-1,4-benzodioxin-6-yl ethanone, 2-bromo-1-2,3-dihydro-1,4-benzodioxin-6-yl ethan-1-one, 2-bromo-1-2,3-dihydrobenzo b 1,4 dioxin-6-yl ethanone, 2-bromo-1-2,3-dihydrobenzo 1,4 dioxin-6-yl ethanone, 2-bromo-1-2,3-dihydrobenzo 1,4-dioxin-6-yl ethanone, 1,4-benzodioxan-6-ylbromomethyl ketone, 2-bromo-1-2,3-dihydro-benzo 1,4 dioxin-6-yl-ethanone, ethanone,2-bromo-1-2,3-dihydro-1,4-benzodioxin-6-yl
BrCC(=O)C1=CC=C2OCCOC2=C1
257.08
257.08
2-Bromo-1-(2, 3-dihydro-1, 4-benzodioxin-6-yl)ethan-1-one
95%
C10H9BrO3
1 g
CSSHRKYOZTZFCX-UHFFFAOYSA-N
2-bromo-1-(2,3-dihydro-1,4-benzodioxin-6-yl)ethanone
2776171
95%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'