CAS RN 4629-54-3
IUPAC Name:
2-bromo-1-(2,3-dihydro-1,4-benzodioxin-6-yl)ethan-1-one
Synonyms:
2-bromo-1-(2,3-dihydro-1,4-benzodioxin-6-yl)ethanone
Molecular Weight (g/mol):
257.08
Molecular Formula:
C10H9BrO3
InChi Key:
CSSHRKYOZTZFCX-UHFFFAOYSA-N
SMILES:
BrCC(=O)C1=CC=C2OCCOC2=C1
1
–
3
of
3
results
1
2-Bromo-1-(2,3-dihydro-1,4-benzodioxin-6-yl)ethan-1-one, Thermo Scientific™
CAS: 4629-54-3 Molecular Formula: C10H9BrO3 Molecular Weight (g/mol): 257.08 MDL Number: MFCD00099402 InChI Key: CSSHRKYOZTZFCX-UHFFFAOYSA-N Synonym: 2-bromo-1-2,3-dihydro-1,4-benzodioxin-6-yl ethanone,2-bromo-1-2,3-dihydro-1,4-benzodioxin-6-yl ethan-1-one,2-bromo-1-2,3-dihydrobenzo b 1,4 dioxin-6-yl ethanone,2-bromo-1-2,3-dihydrobenzo 1,4 dioxin-6-yl ethanone,2-bromo-1-2,3-dihydrobenzo 1,4-dioxin-6-yl ethanone,1,4-benzodioxan-6-ylbromomethyl ketone,2-bromo-1-2,3-dihydro-benzo 1,4 dioxin-6-yl-ethanone,ethanone,2-bromo-1-2,3-dihydro-1,4-benzodioxin-6-yl PubChem CID: 2776171 SMILES: BrCC(=O)C1=CC=C2OCCOC2=C1