missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1,1,2-triphenylethylene 98.0+%, TCI America™

Catalog No. T085325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T085325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T085325G Supplier TCI America Supplier No. T085325G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1607-57-4
Molecular Formula C20H15Br
Molecular Weight (g/mol) 335.244
MDL Number MFCD00000135
InChI Key VUQVJIUBUPPCDB-UHFFFAOYSA-N
Synonym bromotriphenylethylene, eitriphin, triphenylethylene bromide, 2-bromoethene-1,1,2-triyl tribenzene, phenylstilbene bromide, 2-bromo-1,1,2-triphenylethylene, ethylene, bromotriphenyl, 1,1,2-triphenylvinyl bromide, unii-mrw5mko6lz, triphenylvinyl bromide
PubChem CID 15354
IUPAC Name (1-bromo-2,2-diphenylethenyl)benzene
SMILES C1=CC=C(C=C1)C(=C(C2=CC=CC=C2)Br)C3=CC=CC=C3

Specifications

Melting Point 114°C
Color Yellow
Quantity 25 g
Formula Weight 335.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-1,1,2-triphenylethylene
RTECSNumber KU8850000
TSCA No