accessibility menu, dialog, popup

UserName

Br

CAS RN 1607-57-4

IUPAC Name: (1-bromo-2,2-diphenylethenyl)benzene
Synonyms: (1-bromo-2,2-diphenylethenyl)benzene
Molecular Weight (g/mol): 335.24
Molecular Formula: C20H15Br
InChi Key: VUQVJIUBUPPCDB-UHFFFAOYSA-N
SMILES: BrC(=C(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results