missing translation for 'onlineSavingsMsg'
Learn More

2-Biphenylmethanol, 98%

Catalog No. AAL0709406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0709406 5 g
AAL0709403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0709406 Supplier Thermo Scientific Chemicals Supplier No. L0709406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2928-43-0 | C13H12O | 184.238 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2928-43-0
Molecular Formula C13H12O
Molecular Weight (g/mol) 184.238
MDL Number MFCD00004620
InChI Key VKTQADPEPIVMHK-UHFFFAOYSA-N
Synonym 2-biphenylmethanol, 1,1'-biphenyl-2-ylmethanol, 2-phenylphenyl methanol, biphenylmethanol, 2-phenylbenzyl alcohol, 1,1'-biphenyl methanol, biphenyl-2-yl-methanol, 2-phenylphenyl methan-1-ol, 1,1'-biphenyl-2-methanol, biphenyl-2-ylmethanol
PubChem CID 76229
IUPAC Name (2-phenylphenyl)methanol
SMILES C1=CC=C(C=C1)C2=CC=CC=C2CO

Specifications

Melting Point 47°C to 51°C
Boiling Point 96°C (0.04 mmHg)
Quantity 5 g
Beilstein 1866883
Formula Weight 184.24
Percent Purity 98%
Chemical Name or Material 2-Biphenylmethanol
EINECSNumber 220-892-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only