missing translation for 'onlineSavingsMsg'
Learn More

2-Benzylphenol 97.0+%, TCI America™

Catalog No. p-7118360
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B011125G 25 g
B0111500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B011125G Supplier TCI America Supplier No. B011125G
Only null left

Chemical Identifiers

CAS 28994-41-4
Molecular Formula C13H12O
Molecular Weight (g/mol) 184.238
MDL Number MFCD00002242
InChI Key CDMGNVWZXRKJNS-UHFFFAOYSA-N
Synonym 2-hydroxydiphenylmethane, o-benzylphenol, phenol, 2-phenylmethyl, benzylphenol, 2-benzyl-phenol, 2-phenylmethyl phenol, alpha-phenyl-o-cresol, o-cresol, .alpha.-phenyl, unii-sf555bl24o, 2-benzyl phenol
PubChem CID 24216
IUPAC Name 2-benzylphenol
SMILES C1=CC=C(C=C1)CC2=CC=CC=C2O

Specifications

Melting Point 54°C
Boiling Point 312°C
Quantity 25 g
Formula Weight 184.24
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Benzylphenol