missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyloxybenzoic Acid 98.0+%, TCI America™

Catalog No. p-7123734
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47665G 5 g
B476625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47665G Supplier TCI America Supplier No. B47665G
Only null left

Chemical Identifiers

CAS 14389-86-7
Molecular Formula C14H11O3
Molecular Weight (g/mol) 227.24
MDL Number MFCD00051940
InChI Key GMOYUTKNPLBTMT-UHFFFAOYSA-M
Synonym 2-benzyloxy benzoic acid, 2-benzyloxybenzoic acid, benzoic acid,2-phenylmethoxy, 2-phenylmethoxy benzoic acid, benzoic acid, 2-phenylmethoxy, enamine_005339, benzyloxybenzoic acid, acmc-20ao3m, 2-benzyloxy-benzoic acid, 2-benzyloxybenzoicacid
PubChem CID 1810581
IUPAC Name 2-(benzyloxy)benzoate
SMILES [O-]C(=O)C1=CC=CC=C1OCC1=CC=CC=C1

Specifications

Melting Point 76°C
Color White-Yellow
Quantity 5 g
UN Number 3077
Formula Weight 228.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Benzyloxybenzoic Acid