missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyloxy-2-methyl-1-propanol, 95%, Thermo Scientific Chemicals

Catalog No. p-7028614
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2600603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2600603 Supplier Thermo Scientific Chemicals Supplier No. H2600603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 91968-71-7 | Molecular Formula: C11H16O2 | Molecular Weight: 180.25 g/mol

2-Benzyloxy-2-methyl-1-propanol is used as a building block in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Benzyloxy-2-methyl-1-propanol is used as a building block in organic synthesis.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 91968-71-7
Molecular Formula C11H16O2
Molecular Weight (g/mol) 180.25
MDL Number MFCD08459314
InChI Key FRHJVAJQSPVSKV-UHFFFAOYSA-N
Synonym 2-benzyloxy-2-methylpropan-1-ol, 2-benzyloxy-2-methyl-1-propanol, 2-methyl-2-phenylmethoxy propan-1-ol
PubChem CID 42552903
IUPAC Name 2-methyl-2-phenylmethoxypropan-1-ol
SMILES CC(C)(CO)OCC1=CC=CC=C1

Specifications

Boiling Point 80°C to 85°C (0.1 mmHg)
Quantity 1 g
Solubility Information Immiscible with water.
Formula Weight 180.12
Percent Purity 95%
Chemical Name or Material 2-Benzyloxy-2-methyl-1-propanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only