missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyloxy-1,3-propanediol 97.0+%, TCI America™

Catalog No. B22711G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22711G 1 g
B22715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22711G Supplier TCI America Supplier No. B22711G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14690-00-7
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.219
MDL Number MFCD00075493
InChI Key UDIPIOHLDFSMLR-UHFFFAOYSA-N
Synonym 2-O-Benzylglycerol, Glycerol 2-Benzyl Ether
PubChem CID 203567
IUPAC Name 2-phenylmethoxypropane-1,3-diol
SMILES C1=CC=C(C=C1)COC(CO)CO

Specifications

Melting Point 42°C
Quantity 1 g
Formula Weight 182.22
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Benzyloxy-1,3-propanediol
RTECSNumber TY3181000
TSCA No