missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Benzylidenecyclohexanone 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 54°C |
| Boiling Point | 190°C |
| Quantity | 5 g |
| Formula Weight | 186.25 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Benzylidenecyclohexanone |
Safety and Handling
| EINECSNumber | (4)-0242 |
| TSCA | No |