accessibility menu, dialog, popup

UserName

CAS RN 5682-83-7

O(Z)

CAS RN 5682-83-7

IUPAC Name: (2Z)-2-(phenylmethylidene)cyclohexan-1-one
Synonyms: (2Z)-2-(phenylmethylidene)cyclohexan-1-one
Molecular Weight (g/mol): 186.25
Molecular Formula: C13H14O
InChi Key: VCDPHYIZVFJQCD-BENRWUELSA-N
SMILES: O=C1CCCC\C1=C\C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results