missing translation for 'onlineSavingsMsg'
Learn More

2-Benzylidenecyclohexanone 95.0+%, TCI America™

Catalog No. B03851G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B03851G 1 g
B03855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B03851G Supplier TCI America Supplier No. B03851G
Only null left

Chemical Identifiers

CAS 5682-83-7
Molecular Formula C13H14O
Molecular Weight (g/mol) 186.254
MDL Number MFCD00070483
InChI Key VCDPHYIZVFJQCD-BENRWUELSA-N
Synonym 2-benzylidenecyclohexanone, cyclohexanone, 2-benzylidene, cyclohexanone, 2-phenylmethylene, 2-benzylidenecyclohexan-1-one, 2z-2-phenylmethylidene cyclohexan-1-one, z-2-benzylidenecyclohexanone, 2-phenylmethylene cyclohexan-1-one, 2z-2-benzylidenecyclohexanone, 2z-2-benzylidenecyclohexan-1-one
PubChem CID 5383302
IUPAC Name (2Z)-2-benzylidenecyclohexan-1-one
SMILES C1CCC(=O)C(=CC2=CC=CC=C2)C1

Specifications

Melting Point 54°C
Boiling Point 190°C
Quantity 1 g
Formula Weight 186.25
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Benzylidenecyclohexanone
EINECSNumber (4)-0242
TSCA No