missing translation for 'onlineSavingsMsg'
Learn More

2-Benzylcyclohexanone, 97%

Catalog No. p-7044309
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0577603 1 g
AAL0577606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0577603 Supplier Thermo Scientific Chemicals Supplier No. L0577603

CAS: 946-33-8 | C13H16O | 188.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 946-33-8
Molecular Formula C13H16O
Molecular Weight (g/mol) 188.27
MDL Number MFCD00051455
InChI Key CUYLPYBYCYQGCE-UHFFFAOYSA-N
Synonym 2-benzylcyclohexanone, cyclohexanone, 2-phenylmethyl, 2-benzyl cyclohexanone, 2-benzyl-cyclohexan-1-one, 2-phenylmethyl cyclohexanone, cyclohexanone,2-phenylmethyl, 2rs-2-phenylmethylcyclohexanone
PubChem CID 98945
IUPAC Name 2-benzylcyclohexan-1-one
SMILES C1CCC(=O)C(C1)CC2=CC=CC=C2

Specifications

Melting Point 28°C to 30°C
Density 1.03
Boiling Point 103°C to 105°C (0.2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.536
Quantity 1 g
Beilstein 2046670
Formula Weight 188.27
Percent Purity 97%
Chemical Name or Material 2-Benzylcyclohexanone
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-420-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only