missing translation for 'onlineSavingsMsg'
Learn More

2-Benzylcyclohexanone, 97%

Catalog No. aal0577603
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
AAL0577603 1 g
AAL0577606 5 g
2 options

Catalog No. AAL0577603

Supplier: Thermo Scientific Chemicals L0577603

Only null left

CAS: 946-33-8 | C13H16O | 188.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 946-33-8
Molecular Formula C13H16O
Molecular Weight (g/mol) 188.27
MDL Number MFCD00051455
InChI Key CUYLPYBYCYQGCE-UHFFFAOYSA-N
Synonym 2-benzylcyclohexanone, cyclohexanone, 2-phenylmethyl, 2-benzyl cyclohexanone, 2-benzyl-cyclohexan-1-one, 2-phenylmethyl cyclohexanone, cyclohexanone,2-phenylmethyl, 2rs-2-phenylmethylcyclohexanone
PubChem CID 98945
IUPAC Name 2-benzylcyclohexan-1-one
SMILES C1CCC(=O)C(C1)CC2=CC=CC=C2

Specifications

Melting Point 28°C to 30°C
Density 1.03
Boiling Point 103°C to 105°C (0.2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.536
Quantity 1 g
Beilstein 2046670
Formula Weight 188.27
Percent Purity 97%
Chemical Name or Material 2-Benzylcyclohexanone
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-420-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only