Learn More
2-Benzyl-N-Boc-L-proline, 95%
CAS: 706806-60-2 | C17H23NO4 | 305.374 g/mol
Supplier: Thermo Scientific Chemicals H5218403
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Benzyl-N-Boc-L-proline | |
| 154°C to 156°C | |
| MFCD06795522 | |
| boc-r-alpha-benzyl-proline, r-2-benzyl-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, 2-benzyl-1-tert-butoxycarbonyl proline, r-2-benzyl-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester, r-2-benzyl-1-boc-2-pyrrolidinecarboxylic acid, boc-, a-benzyl l-proline, boc-, boc-r-, 2-benzyl-n-boc-l-proline, boc-r-, a-benzyl-pro-oh | |
| CC(C)(C)OC(=O)N1CCCC1(CC2=CC=CC=C2)C(=O)O | |
| 305.374 | |
| 305.37 |
| 706806-60-2 | |
| C17H23NO4 | |
| 1 g | |
| JUUNPRGYFCIXSM-QGZVFWFLSA-N | |
| (2R)-2-benzyl-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid | |
| 2761823 | |
| 95% |
Chemical Identifiers
| 706806-60-2 | |
| 305.374 | |
| JUUNPRGYFCIXSM-QGZVFWFLSA-N | |
| 2761823 | |
| CC(C)(C)OC(=O)N1CCCC1(CC2=CC=CC=C2)C(=O)O |
| C17H23NO4 | |
| MFCD06795522 | |
| boc-r-alpha-benzyl-proline, r-2-benzyl-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, 2-benzyl-1-tert-butoxycarbonyl proline, r-2-benzyl-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester, r-2-benzyl-1-boc-2-pyrrolidinecarboxylic acid, boc-, a-benzyl l-proline, boc-, boc-r-, 2-benzyl-n-boc-l-proline, boc-r-, a-benzyl-pro-oh | |
| (2R)-2-benzyl-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only