missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyl-N-Boc-L-proline, 95%

Catalog No. p-7034498
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52184MD 250 mg
AAH5218403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52184MD Supplier Thermo Scientific Chemicals Supplier No. H52184MD

CAS: 706806-60-2 | C17H23NO4 | 305.374 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 706806-60-2
Molecular Formula C17H23NO4
Molecular Weight (g/mol) 305.374
MDL Number MFCD06795522
InChI Key JUUNPRGYFCIXSM-QGZVFWFLSA-N
Synonym boc-r-alpha-benzyl-proline, r-2-benzyl-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, r-2-benzyl-1-boc-2-pyrrolidinecarboxylic acid, r-2-benzyl-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester, 2r-2-benzyl-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, boc-, a-benzyl l-proline, 2-benzyl-n-boc-l-proline, boc-r-, a-benzyl-pro-oh, boc-r-alpha-benzyl-pro-oh, 2-benzyl-1-tert-butoxycarbonyl proline
PubChem CID 2761823
IUPAC Name (2R)-2-benzyl-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCCC1(CC2=CC=CC=C2)C(=O)O

Specifications

Melting Point 154°C to 156°C
Quantity 250 mg
Formula Weight 305.37
Percent Purity 95%
Chemical Name or Material 2-Benzyl-N-Boc-L-proline
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only