missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyl-L-proline hydrochloride, 95%

Catalog No. p-7034396
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52080MD 250 mg
AAH5208003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52080MD Supplier Thermo Scientific Chemicals Supplier No. H52080MD

CAS: 86116-84-9 | C12H15NO2 | 205.257 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 86116-84-9
Molecular Formula C12H15NO2
Molecular Weight (g/mol) 205.257
MDL Number MFCD16660381
InChI Key LIHLFIASPODZOB-GFCCVEGCSA-N
Synonym 2-benzylproline, 2-benzyl-l-proline, r-2-benzylpyrrolidine-2-carboxylic acid, l-proline, 2-phenylmethyl, r-2-benzyl-pyrrolidine-2-carboxylic acid, r-alpha-benzyl-l-proline-hcl, r-a-benzyl-prolinecl, 2r-2-benzylpyrrolidine-2-carboxylic acid, r-a-benzyl proline?hcl, r-alpha-benzylproline hcl
PubChem CID 2761822
IUPAC Name (2R)-2-benzylpyrrolidine-2-carboxylic acid
SMILES C1CC(NC1)(CC2=CC=CC=C2)C(=O)O

Specifications

Melting Point 270°C to 272°C
Quantity 250 mg
Formula Weight 241.72
Optical Rotation −33° (c=1 in 1N HCl)
Percent Purity 95%
Chemical Name or Material 2-Benzyl-L-proline hydrochloride
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only