missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyl-2-(dimethylamino)-4'-morpholinobutyrophenone 98.0+%, TCI America™

Catalog No. p-7123241
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B391425G 25 g
B3914100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B391425G Supplier TCI America Supplier No. B391425G

Chemical Identifiers

CAS 119313-12-1
Molecular Formula C23H30N2O2
Molecular Weight (g/mol) 366.505
MDL Number MFCD00191775
InChI Key UHFFVFAKEGKNAQ-UHFFFAOYSA-N
Synonym 2-Benzyl-2-(dimethylamino)-1-[4-(morpholino)phenyl]-1-butanone
PubChem CID 86171
IUPAC Name 2-benzyl-2-(dimethylamino)-1-(4-morpholin-4-ylphenyl)butan-1-one
SMILES CCC(CC1=CC=CC=C1)(C(=O)C2=CC=C(C=C2)N3CCOCC3)N(C)C

Specifications

Melting Point 116°C
Color Yellow
Quantity 25 g
UN Number 3077
Formula Weight 366.51
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Benzyl-2-(dimethylamino)-4′-morpholinobutyrophenone
EINECSNumber (5)-6310
RTECSNumber EL7755000
TSCA Yes