missing translation for 'onlineSavingsMsg'
Learn More

2-Benzoylpyrrole 98.0+%, TCI America™

Catalog No. B49345G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49345G 5 g
B493425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49345G Supplier TCI America Supplier No. B49345G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7697-46-3
Molecular Formula C11H9NO
Molecular Weight (g/mol) 171.199
MDL Number MFCD00094048
InChI Key NFGGQMYSOLVBLF-UHFFFAOYSA-N
PubChem CID 24370
IUPAC Name phenyl(1H-pyrrol-2-yl)methanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=CN2

Specifications

Melting Point 77°C
Color White-Yellow
Boiling Point 305°C
Quantity 5 g
Formula Weight 171.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Benzoylpyrrole
RTECSNumber OB6407000
TSCA No