accessibility menu, dialog, popup

UserName

ONH

CAS RN 7697-46-3

IUPAC Name: 2-benzoyl-1H-pyrrole
Synonyms: 2-benzoyl-1H-pyrrole
Molecular Weight (g/mol): 171.2
Molecular Formula: C11H9NO
InChi Key: NFGGQMYSOLVBLF-UHFFFAOYSA-N
SMILES: O=C(C1=CC=CN1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results