missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Benzoylbenzoic Acid 99.0+%, TCI America™
$40.43 - $40.43
Chemical Identifiers
| CAS | 85-52-9 |
|---|---|
| Molecular Formula | C14H10O3 |
| Molecular Weight (g/mol) | 226.23 |
| MDL Number | MFCD00002472 |
| InChI Key | FGTYTUFKXYPTML-UHFFFAOYSA-N |
| Synonym | o-benzoylbenzoic acid, benzoic acid, 2-benzoyl, 2-carboxybenzophenone, benzophenone-2-carboxylic acid, benzoic acid, o-benzoyl, o-carboxybenzophenone, benzophenone-2-carbonic acid, 2-benzoquinonecarboxylic acid, benzoylbenzoic acid, 2-phenylcarbonyl benzoic acid |
| PubChem CID | 6813 |
| IUPAC Name | 2-benzoylbenzoic acid |
| SMILES | OC(=O)C1=CC=CC=C1C(=O)C1=CC=CC=C1 |
Chemical Identifiers
| 85-52-9 | |
| 226.23 | |
| FGTYTUFKXYPTML-UHFFFAOYSA-N | |
| 6813 | |
| OC(=O)C1=CC=CC=C1C(=O)C1=CC=CC=C1 |
| C14H10O3 | |
| MFCD00002472 | |
| o-benzoylbenzoic acid, benzoic acid, 2-benzoyl, 2-carboxybenzophenone, benzophenone-2-carboxylic acid, benzoic acid, o-benzoyl, o-carboxybenzophenone, benzophenone-2-carbonic acid, 2-benzoquinonecarboxylic acid, benzoylbenzoic acid, 2-phenylcarbonyl benzoic acid | |
| 2-benzoylbenzoic acid |
Specifications
| 85-52-9 | |
| White-Yellow | |
| MFCD00002472 | |
| o-benzoylbenzoic acid, benzoic acid, 2-benzoyl, 2-carboxybenzophenone, benzophenone-2-carboxylic acid, benzoic acid, o-benzoyl, o-carboxybenzophenone, benzophenone-2-carbonic acid, 2-benzoquinonecarboxylic acid, benzoylbenzoic acid, 2-phenylcarbonyl benzoic acid | |
| OC(=O)C1=CC=CC=C1C(=O)C1=CC=CC=C1 | |
| 226.23 | |
| 226.23 | |
| Crystalline Powder |
| 128°C | |
| C14H10O3 | |
| 25 g | |
| FGTYTUFKXYPTML-UHFFFAOYSA-N | |
| 2-benzoylbenzoic acid | |
| 6813 | |
| ≥99.0% (T) | |
| 2-Benzoylbenzoic Acid |
Safety and Handling
EINECSNumber : (4)-0268
RTECSNumber : DG3600000
TSCA : Yes