Learn More
2-Benzoylbenzoic acid, 98%
CAS: 85-52-9 | C14H10O3 | 226.23 g/mol
$72.38 - $72.38
Chemical Identifiers
| CAS | 85-52-9 |
|---|---|
| Molecular Formula | C14H10O3 |
| Molecular Weight (g/mol) | 226.23 |
| MDL Number | MFCD00002472 |
| InChI Key | FGTYTUFKXYPTML-UHFFFAOYSA-N |
| Synonym | o-benzoylbenzoic acid, benzoic acid, 2-benzoyl, 2-carboxybenzophenone, benzophenone-2-carboxylic acid, benzoic acid, o-benzoyl, o-carboxybenzophenone, benzophenone-2-carbonic acid, 2-benzoquinonecarboxylic acid, benzoylbenzoic acid, 2-phenylcarbonyl benzoic acid |
| PubChem CID | 6813 |
| IUPAC Name | 2-benzoylbenzoic acid |
| SMILES | OC(=O)C1=CC=CC=C1C(=O)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1328118
|
Thermo Scientific Chemicals
A1328118 |
50 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 85-52-9 | |
| 226.23 | |
| FGTYTUFKXYPTML-UHFFFAOYSA-N | |
| 6813 | |
| OC(=O)C1=CC=CC=C1C(=O)C1=CC=CC=C1 |
| C14H10O3 | |
| MFCD00002472 | |
| o-benzoylbenzoic acid, benzoic acid, 2-benzoyl, 2-carboxybenzophenone, benzophenone-2-carboxylic acid, benzoic acid, o-benzoyl, o-carboxybenzophenone, benzophenone-2-carbonic acid, 2-benzoquinonecarboxylic acid, benzoylbenzoic acid, 2-phenylcarbonyl benzoic acid | |
| 2-benzoylbenzoic acid |
Specifications
| 85-52-9 | |
| 257°C | |
| C14H10O3 | |
| 50 g | |
| o-benzoylbenzoic acid, benzoic acid, 2-benzoyl, 2-carboxybenzophenone, benzophenone-2-carboxylic acid, benzoic acid, o-benzoyl, o-carboxybenzophenone, benzophenone-2-carbonic acid, 2-benzoquinonecarboxylic acid, benzoylbenzoic acid, 2-phenylcarbonyl benzoic acid | |
| OC(=O)C1=CC=CC=C1C(=O)C1=CC=CC=C1 | |
| 226.23 | |
| 226.23 | |
| 2-Benzoylbenzoic acid |
| 127°C to 130°C | |
| Odorless | |
| MFCD00002472 | |
| 1107841 | |
| FGTYTUFKXYPTML-UHFFFAOYSA-N | |
| 2-benzoylbenzoic acid | |
| 6813 | |
| ≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 201-612-2
RTECSNumber : DG3600000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only