missing translation for 'onlineSavingsMsg'
Learn More

(2-Benzothiazolylthio)acetic Acid 98.0+%, TCI America™

Catalog No. b1941500g
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1941500G 500 g
B194125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1941500G Supplier TCI America Supplier No. B1941500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6295-57-4
Molecular Formula C9H7NO2S2
Molecular Weight (g/mol) 225.28
MDL Number MFCD00227391
InChI Key ZZUQWNYNSKJLPI-UHFFFAOYSA-N
PubChem CID 80525
IUPAC Name 2-(1,3-benzothiazol-2-ylsulfanyl)acetic acid
SMILES OC(=O)CSC1=NC2=CC=CC=C2S1

Specifications

Melting Point 159°C
Color Yellow
Quantity 500 g
Formula Weight 225.28
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (2-Benzothiazolylthio)acetic Acid
EINECSNumber (5)-6037
TSCA Yes