missing translation for 'onlineSavingsMsg'
Learn More

2-Benzothiazolyl Diethyldithiocarbamate 95.0+%, TCI America™

Catalog No. D024825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D024825G 25 g
D0248500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D024825G Supplier TCI America Supplier No. D024825G

Chemical Identifiers

CAS 95-30-7
Molecular Formula C12H14N2S3
Molecular Weight (g/mol) 282.438
MDL Number MFCD00059887
InChI Key LFMQNMXVVXHZCC-UHFFFAOYSA-N
Synonym Diethyldithiocarbamic Acid 2-Benzothiazolyl Ester, N,N-Diethylthiocarbamoyl-2-benzothiazolyl Sulfide
PubChem CID 7229
IUPAC Name 1,3-benzothiazol-2-yl N,N-diethylcarbamodithioate
SMILES CCN(CC)C(=S)SC1=NC2=CC=CC=C2S1

Specifications

Melting Point 79°C
Color Yellow
Quantity 25 g
Formula Weight 282.44
Percent Purity ≥95.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Benzothiazolyl Diethyldithiocarbamate
EINECSNumber (5)-0253
RTECSNumber EZ4950000
TSCA Yes