missing translation for 'onlineSavingsMsg'
Learn More

(2-Benzimidazolylthio)acetic acid, 98%

Catalog No. p-7081260
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1122106 5 g
AAA1122114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1122106 Supplier Thermo Scientific Chemicals Supplier No. A1122106

CAS: 3042-00-0 | C9H7N2O2S | 207.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3042-00-0
Molecular Formula C9H7N2O2S
Molecular Weight (g/mol) 207.23
MDL Number MFCD00022673
InChI Key UYNVBLJQBCTRKV-UHFFFAOYSA-M
Synonym 2-benzimidazolythio acetic acid, 2-benzimidazolylthio-acetic acid, 2-benzimidazolylthio acetic acid, 1h-benzoimidazol-2-ylsulfanyl acetic acid, 1h-benzimidazol-2-ylthio acetic acid, 2-1h-benzo d imidazol-2-yl thio acetic acid, 2-1h-1,3-benzodiazol-2-ylsulfanyl acetic acid, 1h-benzimidazol-2-ylsulfanyl acetic acid, 1h-1,3-benzodiazol-2-ylsulfanyl acetic acid, 2-benzimidazol-2-ylthioacetic acid
PubChem CID 137805
IUPAC Name 2-(1H-benzimidazol-2-ylsulfanyl)acetic acid
SMILES [O-]C(=O)CSC1=NC2=CC=CC=C2N1

Specifications

Melting Point ∼215°C (decomposition)
Quantity 5 g
Beilstein 11825
Formula Weight 208.24
Percent Purity 98%
Chemical Name or Material (2-Benzimidazolylthio)acetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only