Learn More
2-Benzimidazoleacetonitrile, 99%
CAS: 4414-88-4 | C9H7N3 | 157.176 g/mol
$149.33 - $549.01
Chemical Identifiers
| CAS | 4414-88-4 |
|---|---|
| Molecular Formula | C9H7N3 |
| Molecular Weight (g/mol) | 157.176 |
| MDL Number | MFCD00005601 |
| InChI Key | BWOVACANEIVHST-UHFFFAOYSA-N |
| Synonym | 2-benzimidazolyl acetonitrile, 2-cyanomethyl benzimidazole, 1h-benzimidazole-2-acetonitrile, 2-benzimidazoleacetonitrile, 2-benzimidazolylacetonitrile, 2-cyanomethylbenzimidazole, 1h-benzimidazol-2-ylacetonitrile, 2-kyanmethylbenzimidazol, benzimidazole, 2-cyanomethyl, 2-1h-1,3-benzodiazol-2-yl acetonitrile |
| PubChem CID | 20455 |
| IUPAC Name | 2-(1H-benzimidazol-2-yl)acetonitrile |
| SMILES | C1=CC=C2C(=C1)NC(=N2)CC#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0082709
|
Thermo Scientific Chemicals
L0082709 |
10 g |
Each for $149.33
|
|
|||||
|
AAL0082718
|
Thermo Scientific Chemicals
L0082718 |
50 g |
Each for $549.01
|
|
|||||
Description
2-Benzimidazoleacetonitrile, is used as a pharmaceutical intermediate. It is also used as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Benzimidazoleacetonitrile, is used as a pharmaceutical intermediate. It is also used as an intermediate in organic synthesis.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Chemical Identifiers
| 4414-88-4 | |
| 157.176 | |
| BWOVACANEIVHST-UHFFFAOYSA-N | |
| 20455 | |
| C1=CC=C2C(=C1)NC(=N2)CC#N |
| C9H7N3 | |
| MFCD00005601 | |
| 2-benzimidazolyl acetonitrile, 2-cyanomethyl benzimidazole, 1h-benzimidazole-2-acetonitrile, 2-benzimidazoleacetonitrile, 2-benzimidazolylacetonitrile, 2-cyanomethylbenzimidazole, 1h-benzimidazol-2-ylacetonitrile, 2-kyanmethylbenzimidazol, benzimidazole, 2-cyanomethyl, 2-1h-1,3-benzodiazol-2-yl acetonitrile | |
| 2-(1H-benzimidazol-2-yl)acetonitrile |
Specifications
| 4414-88-4 | |
| Odorless | |
| MFCD00005601 | |
| 128461 | |
| Insoluble in water. | |
| C1=CC=C2C(=C1)NC(=N2)CC#N | |
| 157.176 | |
| 157.18 | |
| 2-Benzimidazoleacetonitrile |
| 206°C to 211°C | |
| C9H7N3 | |
| 10 g | |
| 2-benzimidazolyl acetonitrile, 2-cyanomethyl benzimidazole, 1h-benzimidazole-2-acetonitrile, 2-benzimidazoleacetonitrile, 2-benzimidazolylacetonitrile, 2-cyanomethylbenzimidazole, 1h-benzimidazol-2-ylacetonitrile, 2-kyanmethylbenzimidazol, benzimidazole, 2-cyanomethyl, 2-1h-1,3-benzodiazol-2-yl acetonitrile | |
| BWOVACANEIVHST-UHFFFAOYSA-N | |
| 2-(1H-benzimidazol-2-yl)acetonitrile | |
| 20455 | |
| 99% |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
GHS P Statement
P280f
Wear protective clothing.
Warning
EINECSNumber : 224-574-9
RTECSNumber : DD5650000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only