Learn More
2-Benzimidazoleacetonitrile, 99%
CAS: 4414-88-4 | C9H7N3 | 157.176 g/mol
Supplier: Thermo Scientific Chemicals L0082709
Description
2-Benzimidazoleacetonitrile, is used as a pharmaceutical intermediate. It is also used as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Benzimidazoleacetonitrile, is used as a pharmaceutical intermediate. It is also used as an intermediate in organic synthesis.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications
| 2-Benzimidazoleacetonitrile | |
| 206°C to 211°C | |
| C9H7N3 | |
| 10 g | |
| 2-benzimidazolyl acetonitrile, 2-cyanomethyl benzimidazole, 1h-benzimidazole-2-acetonitrile, 2-benzimidazoleacetonitrile, 2-benzimidazolylacetonitrile, 2-cyanomethylbenzimidazole, 1h-benzimidazol-2-ylacetonitrile, 2-kyanmethylbenzimidazol, benzimidazole, 2-cyanomethyl, 2-1h-1,3-benzodiazol-2-yl acetonitrile | |
| BWOVACANEIVHST-UHFFFAOYSA-N | |
| 2-(1H-benzimidazol-2-yl)acetonitrile | |
| 20455 | |
| 99% |
| 4414-88-4 | |
| Odorless | |
| MFCD00005601 | |
| 128461 | |
| Insoluble in water. | |
| C1=CC=C2C(=C1)NC(=N2)CC#N | |
| 157.176 | |
| 157.18 |
Chemical Identifiers
| 4414-88-4 | |
| 157.176 | |
| BWOVACANEIVHST-UHFFFAOYSA-N | |
| 20455 | |
| C1=CC=C2C(=C1)NC(=N2)CC#N |
| C9H7N3 | |
| MFCD00005601 | |
| 2-benzimidazolyl acetonitrile, 2-cyanomethyl benzimidazole, 1h-benzimidazole-2-acetonitrile, 2-benzimidazoleacetonitrile, 2-benzimidazolylacetonitrile, 2-cyanomethylbenzimidazole, 1h-benzimidazol-2-ylacetonitrile, 2-kyanmethylbenzimidazol, benzimidazole, 2-cyanomethyl, 2-1h-1,3-benzodiazol-2-yl acetonitrile | |
| 2-(1H-benzimidazol-2-yl)acetonitrile |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
GHS P Statement
P280f
Wear protective clothing.
Warning
EINECSNumber : 224-574-9
RTECSNumber : DD5650000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only