missing translation for 'onlineSavingsMsg'
Learn More

2-Aminothiophenol, 98%

Catalog No. AAA1895214
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1895214 25 g
AAA1895222 100 g
AAA1895236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1895214 Supplier Thermo Scientific Chemicals Supplier No. A1895214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 137-07-5 | C6H7NS | 125.19 g/mol

2-Aminothiophenol was used in the synthesis of 1,5-benzothiazepines derivatives by reacting with 1,3-diphenylpropenone derivatives and using aluminosilicate solid catalysts. It was also used in the synthesis of benzothiazole2 and 3-(benzothiazol-2-yl)coumarin derivatives. The ternary complexes of copper (II) with salicylidene-2-aminothiophenol (L) and glycine, alanine, valine and histidene amino acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminothiophenol was used in the synthesis of 1,5-benzothiazepines derivatives by reacting with 1,3-diphenylpropenone derivatives and using aluminosilicate solid catalysts. It was also used in the synthesis of benzothiazole2 and 3-(benzothiazol-2-yl)coumarin derivatives. The ternary complexes of copper (II) with salicylidene-2-aminothiophenol (L) and glycine, alanine, valine and histidene amino acids.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.

Chemical Identifiers

CAS 137-07-5
Molecular Formula C6H7NS
Molecular Weight (g/mol) 125.19
MDL Number MFCD00007702
InChI Key VRVRGVPWCUEOGV-UHFFFAOYSA-N
Synonym 2-aminothiophenol, o-aminothiophenol, 2-mercaptoaniline, o-mercaptoaniline, o-aminobenzenethiol, 2-mercaptaniline, benzenethiol, 2-amino, 2-amino thiophenol, 1-amino-2-mercaptobenzene, 2-aminobenzene-1-thiol
PubChem CID 8713
IUPAC Name 2-aminobenzenethiol
SMILES NC1=CC=CC=C1S

Specifications

Melting Point 18°C to 23°C
Density 1.17
Boiling Point 234°C
Flash Point 79°C (174°F)
Odor Stench
Refractive Index 1.642
Quantity 25 g
UN Number UN3267
Beilstein 606076
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 125.19
Percent Purity 98%
Chemical Name or Material 2-Aminothiophenol
Show More Show Less
Hazard Category H227-H302-H314-H335
Hazard Statement GHS H Statement
H301-H314-H318-H373-H227-H317
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
May cause an allergic skin reaction.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 205-277-3
RTECSNumber DC0600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only