missing translation for 'onlineSavingsMsg'
Learn More

2-Aminothiophene-3-carboxamide 98.0+%, TCI America™

Catalog No. A31655G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A31655G 5 g
A31651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A31655G Supplier TCI America Supplier No. A31655G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14080-51-4
Molecular Formula C5H6N2OS
Molecular Weight (g/mol) 142.18
MDL Number MFCD00706455
InChI Key WHZIZZOTISTHCT-UHFFFAOYSA-N
PubChem CID 699495
IUPAC Name 2-aminothiophene-3-carboxamide
SMILES NC(=O)C1=C(N)SC=C1

Specifications

Melting Point 150°C
Color White-Yellow
Quantity 5 g
Formula Weight 142.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminothiophene-3-carboxamide