accessibility menu, dialog, popup

UserName

CAS RN 14080-51-4

IUPAC Name: 2-aminothiophene-3-carboxamide
Synonyms: 2-aminothiophene-3-carboxamide
Molecular Weight (g/mol): 142.18
Molecular Formula: C5H6N2OS
InChi Key: WHZIZZOTISTHCT-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(N)SC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results