missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminothiazole 98.0+%, TCI America™
Supplier: TCI America A063325G
Specifications
| 2-Aminothiazole | |
| 90°C | |
| C3H4N2S | |
| 25 g | |
| RAIPHJJURHTUIC-UHFFFAOYSA-N | |
| 1,3-thiazol-2-amine | |
| 2155 | |
| 100.14 | |
| Crystalline Powder |
| 96-50-4 | |
| 143°C | |
| MFCD00005325 | |
| 2-aminothiazole, aminothiazole, 2-thiazolamine, thiazol-2-amine, abadol, 2-thiazolylamine, abadole, basedol, 2-thiazylamine, 4-thiazolin-2-onimine | |
| NC1=NC=CS1 | |
| 100.14 | |
| CHEBI:40782 | |
| ≥98.0% (T) |
Chemical Identifiers
| 96-50-4 | |
| 100.14 | |
| RAIPHJJURHTUIC-UHFFFAOYSA-N | |
| 2155 | |
| 1,3-thiazol-2-amine |
| C3H4N2S | |
| MFCD00005325 | |
| 2-aminothiazole, aminothiazole, 2-thiazolamine, thiazol-2-amine, abadol, 2-thiazolylamine, abadole, basedol, 2-thiazylamine, 4-thiazolin-2-onimine | |
| CHEBI:40782 | |
| NC1=NC=CS1 |
Safety and Handling
EINECSNumber : (9)-0079
RTECSNumber : XJ2100000
TSCA : Yes