missing translation for 'onlineSavingsMsg'
Learn More

2-Aminopyrimidine, 98%

Catalog No. AC104580050
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC104580050 5 g Glass bottle
AC104581000 100 g Plastic bottle
AC104585000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC104580050 Supplier Thermo Scientific Chemicals Supplier No. 104580050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 109-12-6 | C4H5N3 | 95.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109-12-6
Molecular Formula C4H5N3
Molecular Weight (g/mol) 95.11
MDL Number MFCD00006089
InChI Key LJXQPZWIHJMPQQ-UHFFFAOYSA-N
Synonym 2-aminopyrimidine, 2-pyrimidinamine, pyrimidin-2-ylamine, pyrimidinamine, aminopyrimidine, 2-pyrimidiylamine, pyrimidine, 2-amino, 2-pyridiylamine, 1,2-dihydro-2-iminopyrimidine, pyrimidine, amino
PubChem CID 7978
ChEBI CHEBI:38618
IUPAC Name pyrimidin-2-amine
SMILES NC1=NC=CC=N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 124°C to 127°C
Color White to Yellow
Boiling Point 158°C (186.0 Mbar)
Flash Point 103°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Beilstein 24, 80
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetic acid,readily soluble in alcohol,acetone and chloroform,slightly soluble in ether and benzene
Formula Weight 95.1
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Aminopyrimidine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-648-4
RTECSNumber UV6326000
TSCA TSCA

RUO – Research Use Only