accessibility menu, dialog, popup

UserName

NH2NN

CAS RN 109-12-6

IUPAC Name: pyrimidin-2-amine
Synonyms: 2-aminopyrimidine 2-pyridiylamine 2-pyrimidiylamine
Molecular Weight (g/mol): 95.11
Molecular Formula: C4H5N3
InChi Key: LJXQPZWIHJMPQQ-UHFFFAOYSA-N
SMILES: NC1=NC=CC=N1
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (2)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)