missing translation for 'onlineSavingsMsg'
Learn More

2-Aminopyrimidine, 98%

Catalog No. p-7027664
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2459422 100 g
AAB2459436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2459422 Supplier Thermo Scientific Chemicals Supplier No. B2459422

CAS: 109-12-6 | C4H5N3 | 95.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109-12-6
Molecular Formula C4H5N3
Molecular Weight (g/mol) 95.11
MDL Number MFCD00006089
InChI Key LJXQPZWIHJMPQQ-UHFFFAOYSA-N
Synonym 2-aminopyrimidine, 2-pyrimidinamine, pyrimidin-2-ylamine, pyrimidinamine, aminopyrimidine, 2-pyrimidiylamine, pyrimidine, 2-amino, 2-pyridiylamine, 1,2-dihydro-2-iminopyrimidine, pyrimidine, amino
PubChem CID 7978
ChEBI CHEBI:38618
IUPAC Name pyrimidin-2-amine
SMILES NC1=NC=CC=N1

Specifications

Melting Point 123°C to 127°C
Boiling Point 159°C (186 mmHg)
Quantity 100 g
Beilstein 107014
Formula Weight 95.11
Percent Purity 98%
Chemical Name or Material 2-Aminopyrimidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 203-648-4
RTECSNumber UV6326000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only