missing translation for 'onlineSavingsMsg'
Learn More

2-Aminopyridine 99.0+%, TCI America™

Catalog No. A0411500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0411500G 500 g
A041125G 25 g
A0411100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A0411500G Supplier TCI America Supplier No. A0411500G

Chemical Identifiers

CAS 504-29-0
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.117
MDL Number MFCD00006312
InChI Key ICSNLGPSRYBMBD-UHFFFAOYSA-N
Synonym 2-aminopyridine, 2-pyridinamine, 2-pyridylamine, aminopyridine, pyridinamine, o-aminopyridine, 2-aminopyridin, 2-aminopryidine, amino-2 pyridine, pyridine, 2-amino
PubChem CID 10439
IUPAC Name pyridin-2-amine
SMILES C1=CC=NC(=C1)N

Specifications

Melting Point 61°C
Boiling Point 105°C
Quantity 500 g
UN Number 2671
Formula Weight 94.12
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminopyridine
EINECSNumber (5)-0724&(9)-0106
RTECSNumber US1575000
TSCA Yes
Recommended Storage Refrigerator