accessibility menu, dialog, popup

UserName

CAS RN 504-29-0

NH2N

CAS RN 504-29-0

IUPAC Name: pyridin-2-amine
Synonyms: 2-aminopyridine
Molecular Weight (g/mol): 94.12
Molecular Formula: C5H6N2
InChi Key: ICSNLGPSRYBMBD-UHFFFAOYSA-N
SMILES: NC1=CC=CC=N1
Boiling Point 
  • (3)
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (6)
  • (1)
Percent Purity 
  • (3)
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (3)
  • (1)
  • (3)
  • (6)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)