missing translation for 'onlineSavingsMsg'
Learn More

2-Aminopyridine N-Oxide 98.0+%, TCI America™

Catalog No. p-7117052
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0778100MG 100 mg
A07781G 1 g
A07785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A0778100MG Supplier TCI America Supplier No. A0778100MG
Only null left

Chemical Identifiers

CAS 14150-95-9
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD00059168
InChI Key KTPMVZCGIJJWCD-UHFFFAOYSA-N
PubChem CID 139694
IUPAC Name 2-imino-1,2-dihydropyridin-1-ol
SMILES ON1C=CC=CC1=N

Specifications

Melting Point 166°C
Quantity 100 mg
Formula Weight 110.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminopyridine N-Oxide