Learn More
2-Aminopyridine, 99+%
CAS: 504-29-0 | C5H6N2 | 94.12 g/mol
$230.68 - $230.68
Chemical Identifiers
| CAS | 504-29-0 |
|---|---|
| Molecular Formula | C5H6N2 |
| Molecular Weight (g/mol) | 94.12 |
| MDL Number | MFCD00006312 |
| InChI Key | ICSNLGPSRYBMBD-UHFFFAOYSA-N |
| Synonym | 2-aminopyridine, 2-pyridinamine, 2-pyridylamine, aminopyridine, pyridinamine, o-aminopyridine, 2-aminopyridin, 2-aminopryidine, amino-2 pyridine, pyridine, 2-amino |
| PubChem CID | 10439 |
| IUPAC Name | pyridin-2-amine |
| SMILES | C1=CC=NC(=C1)N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC104541000
|
Thermo Scientific Chemicals
104541000 |
100 g | Plastic bottle |
Each for $230.68
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 504-29-0 | |
| 94.12 | |
| ICSNLGPSRYBMBD-UHFFFAOYSA-N | |
| 10439 | |
| C1=CC=NC(=C1)N |
| C5H6N2 | |
| MFCD00006312 | |
| 2-aminopyridine, 2-pyridinamine, 2-pyridylamine, aminopyridine, pyridinamine, o-aminopyridine, 2-aminopyridin, 2-aminopryidine, amino-2 pyridine, pyridine, 2-amino | |
| pyridin-2-amine |
Specifications
| 504-29-0 | |
| 100.0 | |
| Beige-Yellow to Cream | |
| 92°C | |
| 99% min. (GC) | |
| C5H6N2 | |
| MFCD00006312 | |
| 22,428 | |
| 15,469 | |
| Solubility in water: soluble. Other solubilities: soluble in alcohol,benzene,ether and hot,petroleum ether,sparingly soluble in toluene | |
| C1=CC=NC(=C1)N | |
| 94.12 | |
| 94.12 | |
| Crystalline Powder, Crystals or Flakes |
| 99.0 | |
| 55°C to 60°C | |
| 204°C to 210°C | |
| Authentic | |
| Plastic bottle | |
| (C5H4N)NH2 | |
| 100 g | |
| 09,18 | |
| 2-aminopyridine, 2-pyridinamine, 2-pyridylamine, aminopyridine, pyridinamine, o-aminopyridine, 2-aminopyridin, 2-aminopryidine, amino-2 pyridine, pyridine, 2-amino | |
| ICSNLGPSRYBMBD-UHFFFAOYSA-N | |
| pyridin-2-amine | |
| 10439 | |
| 99+% | |
| 2-Aminopyridine, p.a. |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persist
GHS Signal Word: Danger
EINECSNumber : 207-988-4
RTECSNumber : US1575000
TSCA : TSCA
RUO – Research Use Only