missing translation for 'onlineSavingsMsg'
Learn More

2-Aminopyridine-6-methanol, 97%

Catalog No. AAH3348506
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3348506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3348506 Supplier Thermo Scientific Chemicals Supplier No. H3348506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 79651-64-2 | C6H8N2O | 124.143 g/mol

4-Aminopyridine (4-AP), a well-known potassium channel blocker, has been shown to enhance conduction in injured and demyelinated axons. Aminopyridine has a narrow therapeutic range in clinical application. In the current study, we investigated whether AP-MeOH can enhance impulse conduction in an animal model of MS.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Slightly soluble.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 79651-64-2
Molecular Formula C6H8N2O
Molecular Weight (g/mol) 124.143
MDL Number MFCD09863097
InChI Key YGMPMXTWKROXPP-UHFFFAOYSA-N
Synonym 6-aminopyridin-2-yl methanol, 6-amino-2-pyridinemethanol, 2-aminopyridine-6-methanol, 6-amino-pyridin-2-yl-methanol, 2-pyridinemethanol, 6-amino, 2-amino-6-hydroxymethylpyridine, 6-aminopyridine-2-methanol, 6-amino-2-pyridinyl methanol, 2-amino-6-hydroxymethyl pyridine, 2-hydroxymethyl-6-aminopyridine
PubChem CID 11182606
IUPAC Name (6-aminopyridin-2-yl)methanol
SMILES C1=CC(=NC(=C1)N)CO

Specifications

Melting Point 87°C to 92°C
Quantity 5 g
Solubility Information Slightly soluble.
Formula Weight 124.14
Percent Purity 97%
Chemical Name or Material 2-Aminopyridine-6-methanol
Hazard Category H302+H312-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only