missing translation for 'onlineSavingsMsg'
Learn More

(2-Aminopyrid-4-yl)methanol, 97%

Catalog No. AC437370050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC437370050 5 g Glass bottle
AC437370010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC437370050 Supplier Thermo Scientific Chemicals Supplier No. 437370050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 105250-17-7 | C6H9N2O | 125.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 105250-17-7
Molecular Formula C6H9N2O
Molecular Weight (g/mol) 125.15
MDL Number MFCD03791261
InChI Key ZRJJXXDQIQFZBW-UHFFFAOYSA-O
Synonym 2-aminopyridin-4-yl methanol, 2-amino-pyridin-4-yl-methanol, 2-aminopyridine-4-methanol, 2-amino-4-pyridinylmethanol, 4-pyridinemethanol, 2-amino, 2-aminopyridin-4-yl-methanol, 2-amino-4-pyridinemethanol, 2-amino-4-pyridinyl methanol, 2-amino-4-hydroxymethylpyridine, 2-amino-4-pyridyl methan-1-ol
PubChem CID 1515296
IUPAC Name (2-aminopyridin-4-yl)methanol
SMILES NC1=CC(CO)=CC=[NH+]1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 75.5°C to 84.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 124.14
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material (2-Aminopyrid-4-yl)methanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF

RUO – Research Use Only