missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminophenyl Phenyl Sulfide 98.0+%, TCI America™
$70.84 - $197.59
Chemical Identifiers
| CAS | 1134-94-7 |
|---|---|
| Molecular Formula | C12H11NS |
| Molecular Weight (g/mol) | 201.287 |
| InChI Key | DGBISJKLNVVJGD-UHFFFAOYSA-N |
| Synonym | 2-Aminodiphenyl Sulfide, 2-(Phenylthio)aniline |
| PubChem CID | 365393 |
| IUPAC Name | 2-phenylsulfanylaniline |
| SMILES | C1=CC=C(C=C1)SC2=CC=CC=C2N |
Chemical Identifiers
| 1134-94-7 | |
| 201.287 | |
| 2-Aminodiphenyl Sulfide, 2-(Phenylthio)aniline | |
| 2-phenylsulfanylaniline |
| C12H11NS | |
| DGBISJKLNVVJGD-UHFFFAOYSA-N | |
| 365393 | |
| C1=CC=C(C=C1)SC2=CC=CC=C2N |
Specifications
| 1134-94-7 | |
| 258°C | |
| 25 g | |
| 2-Aminodiphenyl Sulfide, 2-(Phenylthio)aniline | |
| C1=CC=C(C=C1)SC2=CC=CC=C2N | |
| 201.287 | |
| 201.29 | |
| Crystalline Powder |
| 43°C | |
| C12H11NS | |
| 3077 | |
| DGBISJKLNVVJGD-UHFFFAOYSA-N | |
| 2-phenylsulfanylaniline | |
| 365393 | |
| ≥98.0% (GC) | |
| 2-Aminophenyl Phenyl Sulfide |
Safety and Handling
EINECSNumber : (3)-4188
TSCA : No