missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminophenol-4-sulfonamide 97.0+%, TCI America™
$74.56 - $596.49
Chemical Identifiers
| CAS | 98-32-8 |
|---|---|
| Molecular Formula | C6H8N2O3S |
| Molecular Weight (g/mol) | 188.20 |
| MDL Number | MFCD00035782 |
| InChI Key | AVQFHKYAVVQYQO-UHFFFAOYSA-N |
| Synonym | 2-aminophenol-4-sulfonamide, 4-hydroxymetanilamide, aminophenol sulfamide, benzenesulfonamide, 3-amino-4-hydroxy, 3-amino-4-hydroxybenzenesulphonamide, o-aminophenol-p-sulfonamide, 3-amino-4-hydroxy-benzenesulfonamide, metanilamide, 4-hydroxy, 2-aminophenol-4-sulfamide, 3-amino-4-hydroxybenzene-1-sulfonamide |
| PubChem CID | 66814 |
| IUPAC Name | 3-amino-4-hydroxybenzene-1-sulfonamide |
| SMILES | NC1=CC(=CC=C1O)S(N)(=O)=O |
Chemical Identifiers
| 98-32-8 | |
| 188.20 | |
| AVQFHKYAVVQYQO-UHFFFAOYSA-N | |
| 66814 | |
| NC1=CC(=CC=C1O)S(N)(=O)=O |
| C6H8N2O3S | |
| MFCD00035782 | |
| 2-aminophenol-4-sulfonamide, 4-hydroxymetanilamide, aminophenol sulfamide, benzenesulfonamide, 3-amino-4-hydroxy, 3-amino-4-hydroxybenzenesulphonamide, o-aminophenol-p-sulfonamide, 3-amino-4-hydroxy-benzenesulfonamide, metanilamide, 4-hydroxy, 2-aminophenol-4-sulfamide, 3-amino-4-hydroxybenzene-1-sulfonamide | |
| 3-amino-4-hydroxybenzene-1-sulfonamide |
Specifications
| 98-32-8 | |
| C6H8N2O3S | |
| 25 g | |
| AVQFHKYAVVQYQO-UHFFFAOYSA-N | |
| 3-amino-4-hydroxybenzene-1-sulfonamide | |
| 66814 | |
| ≥97.0% (HPLC,T) | |
| 2-Aminophenol-4-sulfonamide |
| 199°C | |
| MFCD00035782 | |
| 2-aminophenol-4-sulfonamide, 4-hydroxymetanilamide, aminophenol sulfamide, benzenesulfonamide, 3-amino-4-hydroxy, 3-amino-4-hydroxybenzenesulphonamide, o-aminophenol-p-sulfonamide, 3-amino-4-hydroxy-benzenesulfonamide, metanilamide, 4-hydroxy, 2-aminophenol-4-sulfamide, 3-amino-4-hydroxybenzene-1-sulfonamide | |
| NC1=CC(=CC=C1O)S(N)(=O)=O | |
| 188.20 | |
| 188.20 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-2030
TSCA : Yes