missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminonicotinic Acid 98.0+%, TCI America™
$52.19 - $152.86
Chemical Identifiers
| CAS | 5345-47-1 |
|---|---|
| Molecular Formula | C6H6N2O2 |
| Molecular Weight (g/mol) | 138.13 |
| MDL Number | MFCD00006318 |
| InChI Key | KPIVDNYJNOPGBE-UHFFFAOYSA-N |
| Synonym | 2-aminonicotinic acid, 2-amino-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-amino, 2-amino-nicotinic acid, 2-amino-3-carboxypyridine, nicotinic acid, 2-amino, aminonicotinic acid, h-nh 2 nicot-oh, amino-nicotinic acid, 2-aminonicotinicacid |
| PubChem CID | 72930 |
| ChEBI | CHEBI:68572 |
| IUPAC Name | 2-aminopyridine-3-carboxylic acid |
| SMILES | NC1=NC=CC=C1C(O)=O |
Chemical Identifiers
| 5345-47-1 | |
| 138.13 | |
| KPIVDNYJNOPGBE-UHFFFAOYSA-N | |
| 72930 | |
| 2-aminopyridine-3-carboxylic acid |
| C6H6N2O2 | |
| MFCD00006318 | |
| 2-aminonicotinic acid, 2-amino-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-amino, 2-amino-nicotinic acid, 2-amino-3-carboxypyridine, nicotinic acid, 2-amino, aminonicotinic acid, h-nh 2 nicot-oh, amino-nicotinic acid, 2-aminonicotinicacid | |
| CHEBI:68572 | |
| NC1=NC=CC=C1C(O)=O |
Specifications
| 5345-47-1 | |
| MFCD00006318 | |
| 2-aminonicotinic acid, 2-amino-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-amino, 2-amino-nicotinic acid, 2-amino-3-carboxypyridine, nicotinic acid, 2-amino, aminonicotinic acid, h-nh 2 nicot-oh, amino-nicotinic acid, 2-aminonicotinicacid | |
| NC1=NC=CC=C1C(O)=O | |
| 138.13 | |
| CHEBI:68572 | |
| ≥98.0% (T) | |
| 2-Aminonicotinic Acid |
| C6H6N2O2 | |
| 5 g | |
| KPIVDNYJNOPGBE-UHFFFAOYSA-N | |
| 2-aminopyridine-3-carboxylic acid | |
| 72930 | |
| 138.13 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes