missing translation for 'onlineSavingsMsg'
Learn More

2-Aminonicotinic acid, 98+%

CAS: 5345-47-1 | C6H6N2O2 | 138.13 g/mol

$88.10 - $919.73

Chemical Identifiers

CAS 5345-47-1
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00006318
InChI Key KPIVDNYJNOPGBE-UHFFFAOYSA-N
Synonym 2-aminonicotinic acid, 2-amino-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-amino, 2-amino-nicotinic acid, 2-amino-3-carboxypyridine, nicotinic acid, 2-amino, aminonicotinic acid, h-nh 2 nicot-oh, amino-nicotinic acid, 2-aminonicotinicacid
PubChem CID 72930
ChEBI CHEBI:68572
IUPAC Name 2-aminopyridine-3-carboxylic acid
SMILES NC1=NC=CC=C1C(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1119506
View Documents
Thermo Scientific Chemicals
A1119506
5 g
Each for $88.10
Only null left
 
AAA1119514
View Documents
Thermo Scientific Chemicals
A1119514
25 g
Each for $294.81
Only null left
 
AAA1119522
View Documents
Thermo Scientific Chemicals
A1119522
100 g
Each for $919.73
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5345-47-1
138.13
KPIVDNYJNOPGBE-UHFFFAOYSA-N
72930
2-aminopyridine-3-carboxylic acid
C6H6N2O2
MFCD00006318
2-aminonicotinic acid, 2-amino-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-amino, 2-amino-nicotinic acid, 2-amino-3-carboxypyridine, nicotinic acid, 2-amino, aminonicotinic acid, h-nh 2 nicot-oh, amino-nicotinic acid, 2-aminonicotinicacid
CHEBI:68572
NC1=NC=CC=C1C(O)=O

Specifications

5345-47-1
C6H6N2O2
5 g
2-aminonicotinic acid, 2-amino-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-amino, 2-amino-nicotinic acid, 2-amino-3-carboxypyridine, nicotinic acid, 2-amino, aminonicotinic acid, h-nh 2 nicot-oh, amino-nicotinic acid, 2-aminonicotinicacid
NC1=NC=CC=C1C(O)=O
138.13
CHEBI:68572
≥98%
∼298°C (decomposition)
MFCD00006318
119031
KPIVDNYJNOPGBE-UHFFFAOYSA-N
2-aminopyridine-3-carboxylic acid
72930
138.13
2-Aminonicotinic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 226-296-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only