missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Aminomethyl)thiophene 98.0+%, TCI America™
Supplier: TCI America A241925G
Specifications
| 2-(Aminomethyl)thiophene | |
| Yellow | |
| C5H7NS | |
| 25 g | |
| 2-thiophenemethylamine, 2-thienylmethylamine, 2-thiophenemethanamine, 2-aminomethyl thiophene, thiophene-2-methylamine, thiophenemethanamine, 2-aminomethylthiophene, 2-thenylamine, 2-thienylmethyl amine, 1-thiophen-2-yl methanamine | |
| C1=CSC(=C1)CN | |
| 113.178 | |
| 113.18 | |
| Liquid |
| 27757-85-3 | |
| 99°C | |
| MFCD00005460 | |
| 2735 | |
| FKKJJPMGAWGYPN-UHFFFAOYSA-N | |
| thiophen-2-ylmethanamine | |
| 34005 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 27757-85-3 | |
| 113.178 | |
| FKKJJPMGAWGYPN-UHFFFAOYSA-N | |
| 34005 | |
| C1=CSC(=C1)CN |
| C5H7NS | |
| MFCD00005460 | |
| 2-thiophenemethylamine, 2-thienylmethylamine, 2-thiophenemethanamine, 2-aminomethyl thiophene, thiophene-2-methylamine, thiophenemethanamine, 2-aminomethylthiophene, 2-thenylamine, 2-thienylmethyl amine, 1-thiophen-2-yl methanamine | |
| thiophen-2-ylmethanamine |
Safety and Handling
TSCA : No