missing translation for 'onlineSavingsMsg'
Learn More

2-(Aminomethyl)thiophene 98.0+%, TCI America™

Catalog No. p-7117895
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24195G 5 g
A241925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24195G Supplier TCI America Supplier No. A24195G
Only null left

Chemical Identifiers

CAS 27757-85-3
Molecular Formula C5H7NS
Molecular Weight (g/mol) 113.178
MDL Number MFCD00005460
InChI Key FKKJJPMGAWGYPN-UHFFFAOYSA-N
Synonym 2-thiophenemethylamine, 2-thienylmethylamine, 2-thiophenemethanamine, 2-aminomethyl thiophene, thiophene-2-methylamine, thiophenemethanamine, 2-aminomethylthiophene, 2-thenylamine, 2-thienylmethyl amine, 1-thiophen-2-yl methanamine
PubChem CID 34005
IUPAC Name thiophen-2-ylmethanamine
SMILES C1=CSC(=C1)CN

Specifications

Color Yellow
Boiling Point 99°C
Quantity 5 g
UN Number 2735
Formula Weight 113.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(Aminomethyl)thiophene