missing translation for 'onlineSavingsMsg'
Learn More

2-(Aminomethyl)benzimidazole dihydrochloride hydrate, 98%

Catalog No. AAL0744006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0744006 5 g
AAL0744014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0744006 Supplier Thermo Scientific Chemicals Supplier No. L0744006

CAS: 5993-91-9 | C8H11Cl2N3 | 220.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5993-91-9
Molecular Formula C8H11Cl2N3
Molecular Weight (g/mol) 220.10
MDL Number MFCD00012677
InChI Key HAEYZZSSSUIZAN-UHFFFAOYSA-N
Synonym 1h-benzo d imidazol-2-yl methanamine dihydrochloride, 2-aminomethyl benzimidazole dihydrochloride, 1h-benzimidazole-2-methanamine, dihydrochloride, 1h-1,3-benzodiazol-2-ylmethanamine dihydrochloride, 1h-1,3-benzodiazol-2-yl methanamine dihydrochloride, 2-aminomethylbenzimidazole dihydrochloride, c-1h-benzoimidazol-2-yl-methylamine dihydrochloride, benzimidazol-2-ylmethylamine, chloride, chloride, 1-1h-1,3-benzodiazol-2-yl methanamine dihydrochloride, 1h-benzo d imidazol-2-yl methanamine 2hcl
PubChem CID 2723957
IUPAC Name 1H-benzimidazol-2-ylmethanamine;dihydrochloride
SMILES Cl.Cl.NCC1=NC2=CC=CC=C2N1

Specifications

Melting Point ∽264°C (decomposition)
Quantity 5 g
Beilstein 3698942
Formula Weight 220.10 (Anhydrous)
Percent Purity 98%
Chemical Name or Material 2-(Aminomethyl)benzimidazole dihydrochloride hydrate
EINECSNumber 227-820-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only