Learn More
2-Aminomethyl-1H-imidazole dihydrochloride, 97%
CAS: 22600-77-7 | C4H9Cl2N3 | 170.04 g/mol
$567.42 - $567.42
Chemical Identifiers
| CAS | 22600-77-7 |
|---|---|
| Molecular Formula | C4H9Cl2N3 |
| Molecular Weight (g/mol) | 170.04 |
| MDL Number | MFCD06738779 |
| InChI Key | KYUDBQDDNKPSIC-UHFFFAOYSA-N |
| Synonym | 1h-imidazol-2-yl methanamine dihydrochloride, 2-aminomethylimidazole dihydrochloride, 2-aminomethyl-1h-imidazole dihydrochloride, 1h-imidazol-2-ylmethylamine dihydrochloride, 1h-imidazol-2-ylmethanamine dihydrochloride, 1h-imidazole-2-methanamine dihydrochloride, 1h-imidazole-2-methanamine, dihydrochloride, 1-1h-imidazol-2-yl methanamine dihydrochloride, 1h-imidazol-2-ylmethyl amine dihydrochloride, 2-aminomethylimidazole hcl |
| PubChem CID | 12417863 |
| SMILES | Cl.Cl.NCC1=NC=CN1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC454590050
|
Thermo Scientific Chemicals
454590050 |
5 g |
Each for $567.42
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 22600-77-7 | |
| 170.04 | |
| KYUDBQDDNKPSIC-UHFFFAOYSA-N | |
| 12417863 |
| C4H9Cl2N3 | |
| MFCD06738779 | |
| 1h-imidazol-2-yl methanamine dihydrochloride, 2-aminomethylimidazole dihydrochloride, 2-aminomethyl-1h-imidazole dihydrochloride, 1h-imidazol-2-ylmethylamine dihydrochloride, 1h-imidazol-2-ylmethanamine dihydrochloride, 1h-imidazole-2-methanamine dihydrochloride, 1h-imidazole-2-methanamine, dihydrochloride, 1-1h-imidazol-2-yl methanamine dihydrochloride, 1h-imidazol-2-ylmethyl amine dihydrochloride, 2-aminomethylimidazole hcl | |
| Cl.Cl.NCC1=NC=CN1 |
Specifications
| 22600-77-7 | |
| Authentic | |
| C4H9Cl2N3 | |
| 5 g | |
| KYUDBQDDNKPSIC-UHFFFAOYSA-N | |
| dihydrogen 1-(1H-imidazol-2-yl)methanamine dichloride | |
| 12417863 | |
| 98% |
| 250.0°C to 251.0°C | |
| 97.5 to max. 102.5% min. (Argentometry) | |
| MFCD06738779 | |
| 1h-imidazol-2-yl methanamine dihydrochloride, 2-aminomethylimidazole dihydrochloride, 2-aminomethyl-1h-imidazole dihydrochloride, 1h-imidazol-2-ylmethylamine dihydrochloride, 1h-imidazol-2-ylmethanamine dihydrochloride, 1h-imidazole-2-methanamine dihydrochloride, 1h-imidazole-2-methanamine, dihydrochloride, 1-1h-imidazol-2-yl methanamine dihydrochloride, 1h-imidazol-2-ylmethyl amine dihydrochloride, 2-aminomethylimidazole hcl | |
| Cl.Cl.NCC1=NC=CN1 | |
| 170.04 | |
| 170.04 | |
| 2-Aminomethyl-1H-imidazole dihydrochloride |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
RUO – Research Use Only