accessibility menu, dialog, popup

UserName

CAS RN 22600-77-7

IUPAC Name: 1-(1H-imidazol-2-yl)methanamine dihydrochloride
Synonyms: 1-(1H-imidazol-2-yl)methanamine dihydrochloride
Molecular Weight (g/mol): 170.04
Molecular Formula: C4H9Cl2N3
InChi Key: KYUDBQDDNKPSIC-UHFFFAOYSA-N
SMILES: Cl.Cl.NCC1=NC=CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results