missing translation for 'onlineSavingsMsg'
Learn More

2-(Aminomethyl)-1-ethylpyrrolidine 98.0+%, TCI America™

Catalog No. A25565G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25565G 5 g
A255625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25565G Supplier TCI America Supplier No. A25565G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 26116-12-1
Molecular Formula C7H16N2
Molecular Weight (g/mol) 128.219
MDL Number MFCD00003178
InChI Key UNRBEYYLYRXYCG-UHFFFAOYSA-N
Synonym 2-aminomethyl-1-ethylpyrrolidine, 1-ethylpyrrolidin-2-yl methanamine, n-ethyl-2-aminomethylpyrrolidine, 2-pyrrolidinemethanamine, 1-ethyl, 1-ethylpyrrolidin-2-ylmethylamine, 1-ethylpyrrolidin-2-yl methylamine, 1-ethyl-2-aminomethylpyrrolidine, 1-ethyl-2-aminomethyl pyrrolidine, 1-1-ethylpyrrolidin-2-yl methanamine
PubChem CID 117295
IUPAC Name (1-ethylpyrrolidin-2-yl)methanamine
SMILES CCN1CCCC1CN

Specifications

Color Yellow
Boiling Point 60°C
Quantity 5 g
UN Number 2733
Formula Weight 128.22
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(Aminomethyl)-1-ethylpyrrolidine
EINECSNumber (9)-0211
TSCA No